Learn More
Biomatik Corporation Human Filamin C Gamma (FLNC) ELISA Kit
Supplier:  Biomatik Corporation EKU04226
Encompass_Preferred
Human Filamin C Gamma (FLNC) ELISA Kit for detecting FLNC in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.115ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q14315. Gene ID: 2318. Target Synonyms: ABP-280; ABP280A; ABPA; ABPL; FLN2; Actin Binding Protein 280; ABP-280-like protein; Actin-binding-like protein; Filamin-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8975