Learn More
BIOHIPPO Human FKBP1B Microsample
SDP
Catalog No. 5000314905
Encompass
Human FKBP1B Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of FKBP1B in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 056 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human FKBP1B Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human FKBP1B Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human FKBP1B biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14905 Supplier BIOHIPPO Supplier No. BHE15211402
Human FKBP1B Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of FKBP1B in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 056 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human FKBP1B Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human FKBP1B Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human FKBP1B biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color