Learn More
Biomatik Corporation Human Flavin Containing Monooxygenase 1 (FMO1) ELISA Kit
Supplier:  Biomatik Corporation EKU04236
Encompass_Preferred
Human Flavin Containing Monooxygenase 1 (FMO1) ELISA Kit for detecting FMO1 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.115ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q01740. Gene ID: 2326. Target Synonyms: Dimethylaniline oxidase 1; Fetal hepatic flavin-containing monooxygenase 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6614