Learn More
BIOHIPPO Human FLNC Microsample
SDP
Catalog No. 5000314289
Encompass
Human FLNC Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of FLNC in Serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 119 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human FLNC Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human FLNC Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human FLNC biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14289 Supplier BIOHIPPO Supplier No. BHE15211839
Human FLNC Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of FLNC in Serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 119 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human FLNC Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human FLNC Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human FLNC biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color