Learn More
Biomatik Corporation Human Flotillin 2 (FLOT2) ELISA Kit
Supplier:  Biomatik Corporation EKU04238
Encompass_Preferred
Human Flotillin 2 (FLOT2) ELISA Kit for detecting FLOT2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.217ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q14254. Gene ID: 2319. Target Synonyms: ECS1; ESA; ESA1; M17S1; Epidermal Surface Antigen 1; Membrane Component,Chromosome 17,Surface Marker 1; 35kD Protein Identified By Monoclonal Antibody ECS-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8973