Learn More
ACROBiosystems ACROBiosystems Human Flt-3 / Flk-2 Protein, His Tag (MALS verified)
Catalog No. 502106265
Encompass_Preferred
Recombinant Protein;100UG;Human Flt-3, His Tag (FL3-H52H6) is expressed from human 293 cells (HEK293). It contains AA Asn 27 - Ser 543 (Accession # P36888-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 60.4 kDa. The protein migrates as 70-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized ActiveMax Human Flt-3 Ligand, Tag Free (Cat. No. FLL-H5218) at 10 μg/mL (100 μL/well) can bind Human Flt-3, His Tag (Cat. No. FL3-H52H6) with a linear range of 0.078-1.25 μg/mL (QC tested).
Catalog No. 50-210-6265 Supplier ACROBiosystems Supplier No. FL3H52H6100UG
Recombinant Protein;100UG;Human Flt-3, His Tag (FL3-H52H6) is expressed from human 293 cells (HEK293). It contains AA Asn 27 - Ser 543 (Accession # P36888-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 60.4 kDa. The protein migrates as 70-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized ActiveMax Human Flt-3 Ligand, Tag Free (Cat. No. FLL-H5218) at 10 μg/mL (100 μL/well) can bind Human Flt-3, His Tag (Cat. No. FL3-H52H6) with a linear range of 0.078-1.25 μg/mL (QC tested).