Learn More
Sino Biological Human FLT1 MAb 25TESTS 10136-R111-F-25
Supplier:  Sino Biological 10136R111F25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human VEGFR1 / FLT-1 (rh VEGFR1 / FLT-1; Catalog#10136-H08H; P17948-1; Met 1-Asn 756) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-240-8807