Learn More
Sino Biological Recombinant Human VEGFR1/FLT1 Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 10136H08H1
Encompass_Preferred
A DNA sequence encoding the human VEGFR1 (P17948-1) extracellular domain (Met1-Asn756) was expressed fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-5455