Learn More
FUJIFILM BIOSCIENCES INC Human Flt3-Ligand , Recombinant Protein, Shenandoah Biotechnology, 100 ug vial
Supplier:  FUJIFILM BIOSCIENCES INC 10021100UG
Encompass_Preferred
Fms-related tyrosine kinase 3 ligand (FLT-3 ligand) is a growth factor that regulates hematopoietic cell proliferation. FLT-3 ligand signalling is transmitted through the fms-related tyrosine kinase 3 (FLT-3) receptor. FLT-3 ligand promotes the long-term expansion and differentiation of pro-B cells in the presence of interleukin 7 (IL-7) or in combination of IL-7 and interleukin 3 (IL-3). Human FLT-3 ligand is active on mouse cells.
Catalog No. 50-990-722
Edge