Learn More
Sino Biological Anti-FLT3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 310683T08100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human FLT3 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-241-1035