Learn More
Sino Biological Recombinant Anti-VEGFR3/FLT4 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10806R005100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human VEGFR3 / FLT-4 (rh VEGFR3 / FLT-4 ; Catalog#10806-H08H; NP_002011.2; Met1-Ile776).
Catalog No. 50-241-2546