Learn More
Sino Biological Anti-VEGFR3/FLT4 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10806MM0220
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human VEGFR3 extracellular domain (rhVEGFR3; Catalog#10806-H08H; Met1-Ile776; NP_002011.2). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-1381