Learn More
Sino Biological Human FLT4 Protein 20ug (Sino Biological)
Supplier:  Sino Biological 10806H27HB20
Encompass_Preferred
A DNA sequence encoding the human VEGFR3/FLT4 (NP_002011.2) (Met1-Ile776) was expressed with a C-terminal Avi tag followed by a His tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-237-1818