Learn More
Discovery Life Sciences HUMAN FM05
SDP
Catalog No. NC9941971
Encompass
Human FMO5 SUPERSOMES 2.5 mg protein in 0.5mL This activity is catalyzed by FMO5 which is expressed from human FMO5 cDNA using a baculovirus expression system
Catalog No. NC9941971 Supplier Discovery Life Sciences Supplier No. 456245
Human FMO5 SUPERSOMES 2.5 mg protein in 0.5mL This activity is catalyzed by FMO5 which is expressed from human FMO5 cDNA using a baculovirus expression system