Learn More
Discovery Life Sciences HUMAN FMO1 0.5ML@5MG/ML
SDP
Catalog No. NC9828362
Encompass
Human FMO1 SUPERSOMES;Available size: 0.5 ml. Total protein concentration: 5.0 mg/ml. Catalytic assay: methyl p-tolyl sulfide oxidase activity, representative catalytic activity: 10,000 pmol product/(min x mg protein).
Catalog No. NC9828362 Supplier Discovery Life Sciences Supplier No. 456241
Human FMO1 SUPERSOMES;Available size: 0.5 ml. Total protein concentration: 5.0 mg/ml. Catalytic assay: methyl p-tolyl sulfide oxidase activity, representative catalytic activity: 10,000 pmol product/(min x mg protein).