Learn More
Discovery Life Sciences HUMAN FMO3 0.5ML@5MG/ML
SDP
Catalog No. NC9812713
Encompass
Human FMO3 SUPERSOMES, Available size: 2.5 mg protein in 0.5 ml. Catalytic assay: methyl p-tolyl sulfide oxidase, representative catalytic activity: 30,000 pmole/(mg x min).Protein Content: 5.0 mg/ml in 100 mM potassium phosphate (pH 7.4)
Catalog No. NC9812713 Supplier Discovery Life Sciences Supplier No. 456233
Human FMO3 SUPERSOMES, Available size: 2.5 mg protein in 0.5 ml. Catalytic assay: methyl p-tolyl sulfide oxidase, representative catalytic activity: 30,000 pmole/(mg x min).Protein Content: 5.0 mg/ml in 100 mM potassium phosphate (pH 7.4)