Learn More
BIOHIPPO Human FMOD Microsample
SDP
Catalog No. 5000314142
Encompass
Human FMOD Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of FMOD in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 14 5 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human FMOD Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human FMOD Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human FMOD biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14142 Supplier BIOHIPPO Supplier No. BHE15212138
Human FMOD Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of FMOD in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 14 5 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human FMOD Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human FMOD Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human FMOD biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color