Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human FMRP (aa 550-632) Control Fragment Recombinant Protein

Catalog No. RP103064
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP103064 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP103064 Supplier Invitrogen™ Supplier No. RP103064
  • $283.00
    $212.65 / Each of 1

    Save $70.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (89%), Rat (89%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

FMR1 binds RNA and is associated with polysomes. The protein may be involved in mRNA trafficking from the nucleus to the cytoplasm. A trinucleotide repeat in the 5' UTR is normally found at 6-53 copies, but an expansion to 55-230 repeats is the cause of fragile X syndrome. Expansion of the trinucleotide repeat may also cause one form of premature ovarian failure. Multiple alternatively spliced transcript variants that encode different protein isoforms and which are located in different cellular locations have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q06787
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 2332
Name Human FMRP (aa 550-632) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias AT24755; BcDNA:GM08679; cg 6203 gene product from transcript cg6203-rc; cg6203; CG6203-PA; CG6203-PB; CG6203-PC; CG6203-PD; CG6203-PE; CG6203-PF; CG6203-PG; CG6203-PH; CG6203-PI; CG6203-PJ; CG6203-PK; dFMR; DFmr1; dFmrp; dfxr; dfxr1; dFXRP; Dmel\CG6203; Dmel_CG6203; dmfr1; drosophila fragile X mental retardation protein; EP(3)3517; FMR; FMR1; Fmr-1; Fmr1-PA; Fmr1-PB; Fmr1-PC; Fmr1-PD; Fmr1-PE; Fmr1-PF; Fmr1-PG; Fmr1-PH; Fmr1-PI; Fmr1-PJ; Fmr1-PK; FMRP; FMRP translational regulator 1; fragile X; fragile X mental retardation; fragile X mental retardation 1; fragile X mental retardation gene; fragile X mental retardation protein; fragile X mental retardation protein 1; Fragile X mental retardation protein 1 homolog; fragile X mental retardation related 1; fragile X mental retardation syndrome 1; fragile X mental retardation syndrome 1 homolog; Fragile X mental retardation syndrome-related protein 1; fragile X mental retardation-1 protein; fragile X protein; fragile x related; fragile X related protein; fragile X retardation 1 protein; fragile X-related; fragile-X; Fragile-X mental retardation 1; Fragile-X mental retardation protein; Fragile-X-related; FRAXA; FXR; MGC87458; POF; POF1; protein FMR-1; ragile X mental retardation protein; synaptic functional regulator FMR1
Common Name FMRP
Gene Symbol FMR1
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence FKGNDDHSRTDNRPRNPREAKGRTTDGSLQIRVDCNNERSVHTKTLQNTSSEGSRLRTGKDRNQKKEKPDSVDGQQPLVNGVP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.