Learn More
Abcam Human FMRP ELISA Kit, Sandwich ELISA, 96 wells, Colorimetric detection, Sensitive to 99.67 pg/mL, Detection range 390.63 - 25000 pg/mL, Assay time 90 min
Supplier:  Abcam AB2248851X96TESTS
Encompass_Preferred
The Abcam Human FMRP ELISA Kit (ab224885) is a rapid, single-wash 90-minute assay optimized for the quantitative measurement of Human FMRP in cell culture extracts and cell lysates. This kit is designed to provide high sensitivity and a streamlined protocol for accurate detection of Fragile X Mental Retardation Protein (FMRP) suitable for a variety of research applications.
  • Utilizes a SimpleStep ELISA approach for ease of use
  • High sensitivity of 99.67 pg/mL
  • Wide detection range from 390.63 to 25000 pg/mL
  • Quick assay time of 1 hour 30 minutes
  • Single wash step minimizes complexity
  • Colorimetric detection method
  • Compatible with standard laboratory equipment
  • Designed for use with cell culture extracts and cell lysates
  • Intended for research use only
  • Store at +4°C, with a shelf life of 1 year
Catalog No. NC2477280