Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human FN1 native Protein from human plasma

Catalog No. 89944968 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-944-968 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-944-968 Supplier Abnova™ Supplier No. P4955
Add to Cart
Add to Cart

Used for SDS-PAGE

This gene encodes fibronectin, a glycoprotein present in a soluble dimeric form in plasma, and in a dimeric or multimeric form at the cell surface and in extracellular matrix. Fibronectin is involved in cell adhesion and migration processes including embryogenesis, wound healing, blood coagulation, host defense, and metastasis. The gene has three regions subject to alternative splicing, with the potential to produce 20 different transcript variants. However, the full-length nature of some variants has not been determined. [provided by RefSeq]

Specifications

Concentration 22.0 mg/mL
For Use With (Application) SDS-PAGE
Formulation In 0.1M Tris-HCl, 0.15M NaCl, pH 7.4
Gene ID (Entrez) 2335
Molecular Weight (g/mol) 220kDa
Name FN1 (Human) Native Protein
Preparation Method Native protein purified from human plasma
Purification Method Column chromatography
Quantity 1 mg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CIG/DKFZp686F10164/DKFZp686H0342/DKFZp686I1370/DKFZp686O13149/ED-B/FINC/FN/FNZ/GFND/GFND2/LETS/MSF
Common Name FN1
Gene Symbol FN1
Species Human
Protein Tag None
Expression System Native protein purified from human plasma
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.