Learn More
Sino Biological Recombinant Human Fibronectin Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10314H08H2100
Encompass_Preferred
A DNA sequence encoding the human FN1 (NP_997647.1) (Asn1722-Thr1811) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-5384