Learn More
Biomatik Corporation Human Folate Receptor 1, Adult (FOLR1) ELISA Kit
Supplier:  Biomatik Corporation EKU04244
Encompass_Preferred
Human Folate Receptor 1, Adult (FOLR1) ELISA Kit for detecting FOLR1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.6pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P15328. Gene ID: 2348. Target Synonyms: FBP; FOLR; FR-alpha; MOv18; Adult folate-binding protein; KB cells FBP; Ovarian tumor-associated antigen MOv18. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5605