Learn More
Sino Biological Anti-PSMA/FOLH1 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 15877MM02100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PSMA / FOLH1 (rh PSMA / FOLH1; Catalog#15877-H07H; NP_004467.1; Lys44-Ala750). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-4082