Learn More
Sino Biological Recombinant Human PSMA/FOLH1 Protein (ECD, His & AVI Tag), Biotinylated
Supplier:  Sino Biological 15877H56HB100
Encompass_Preferred
A DNA sequence encoding the Human FOLH1 (NP_004467.1) (Lys44-Ala750) was expressed with a N-terminal polyhistidine tag followed by an AVI tag.The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-241-1721