Learn More
Biomatik Corporation Human Follicle Stimulating Hormone (FSH) ELISA Kit
Supplier:  Biomatik Corporation EKL57356
Encompass_Preferred
Human Follicle Stimulating Hormone (FSH) ELISA Kit for detecting FSH in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40miu/ml. Sensitivity: 0.298miu/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 3h. Accession Number: P01225. Gene ID: 2488. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5320