Learn More
Biomatik Corporation Human Follicle Stimulating Hormone (FSH) Instant ELISA Kit
Supplier:  Biomatik Corporation EKU10340
Encompass_Preferred
Human Follicle Stimulating Hormone (FSH) Instant ELISA Kit for detecting FSH in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50mIU/ml. Sensitivity: 0.31mIU/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 1h, 10min. Accession Number: P01215. Gene ID: 1081. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8344