Learn More
Biomatik Corporation Human Folliculin (FLCN) ELISA Kit
Supplier:  Biomatik Corporation EKU04252
Encompass_Preferred
Human Folliculin (FLCN) ELISA Kit for detecting FLCN in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.112ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8NFG4. Gene ID: 201163. Target Synonyms: BHD; FLCL; BHD skin lesion fibrofolliculoma protein; Birt-Hogg-Dube syndrome protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5544