Learn More
ACROBiosystems APC-Labeled Human FOLR1 Protein, His TagStar Staining
Catalog No. 502331449
Encompass_Preferred
APC-Labeled Human FOLR1 Protein, His Tag (FO1-HA2H8) is produced via conjugation of APC to Human FOLR1 Protein, His Tag with a new generation site-specific technology under Star Staining labeling platform. Human FOLR1 Protein, His Tag is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Met 233 (Accession # P15328-1).
Catalog No. 50-233-1449 Supplier ACROBiosystems Supplier No. FO1HA2H825TESTS
APC-Labeled Human FOLR1 Protein, His Tag (FO1-HA2H8) is produced via conjugation of APC to Human FOLR1 Protein, His Tag with a new generation site-specific technology under Star Staining labeling platform. Human FOLR1 Protein, His Tag is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Met 233 (Accession # P15328-1).