Learn More
Raybiotech Inc Human FOLR1 ELISA Kit
Supplier:  Raybiotech Inc ELHFOLR11
Encompass_Preferred
For serum, plasma, and cell culture supernatant samples. Sensitivity: 0.10 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: Cross Reactivity: This ELISA kit shows no cross-reactivity with the following cytokines tested: human ACE-2, aFGF (FGF-1), Albumin, AMICA, Angiopoietin-4, BAFF, CA19-9, CD163, Clusterin, CRTAM, CXCL14 (BRAK), Cystatin C, Decorin, Dkk-3, DLL1, Fetuin A, Furin, GASP-1, GASP-2, GCSF R (CD114), HAI-2, IL-17 RB, IL-27, LAG-3, LDL R, Pepsinogen 1, RANK, RBP4, SOST, Syndecan-1, TACI, TFPI, Thrombospondin 1, TRAIL R2, TRANCE, Troponin I, uPA, VE-Cadherin (CDH5), WISP-1.
Catalog No. 50-194-9276