Learn More
Sino Biological Anti-FOLR1 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 11241MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human FOLR1 (rh FOLR1; Catalog#11241-H08H; NP_057937.1; Met1-Met233). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-1301