Learn More
Sino Biological Recombinant Human FOLR1 Protein (ECD, hFc & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 11241H41HB100
Encompass_Preferred
A DNA sequence encoding the Human FOLR1 (NP_057937.1)(Met1-Met233) was expressed with a c-terminal AVI tagged Fc region of human IgG1 at the C-terminus (Fc-AVI). The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-241-1274