Learn More
Abcam Human FOLR3 Antibody Pair - BSA and Azide free, 10 x 96 Tests
Catalog No. 502691671
Encompass_Preferred
Human FOLR3 Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human FOLR3.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-269-1671 Supplier Abcam Supplier No. AB30370310X96TESTS
Human FOLR3 Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human FOLR3.
The product is subject to the following: Abcam Restricted Use Statement