Learn More
ACROBiosystems ACROBiosystems Human FOLR3 Protein, His Tag
Supplier:  ACROBiosystems FO3H52H3100UG
Encompass_Preferred
Recombinant Protein;100UG;Human FOLR3, His Tag (FO3-H52H3) is expressed from human 293 cells (HEK293). It contains AA Gln 23 - Ser 245 (Accession # P41439-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 27.4 kDa.
Catalog No. 50-210-6865