Learn More
Biomatik Corporation Human Formin Binding Protein 1 (FNBP1) ELISA Kit
Supplier:  Biomatik Corporation EKU04265
Encompass_Preferred
Human Formin Binding Protein 1 (FNBP1) ELISA Kit for detecting FNBP1 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q96RU3. Gene ID: 23048. Target Synonyms: FBP17. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6031