Learn More
Raybiotech Inc Human FoxO3/FKHRL1 ELISA Kit
Supplier:  Raybiotech Inc ELHFOXO31
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.14 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human FoxO3.
Catalog No. 50-194-9279