Learn More
Sino Biological Human FOXP3 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 110647MM02100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, a synthetic peptide corresponding to the center region of the human FOXP3. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-2600