Learn More
ACROBiosystems ACROBiosystems Human Frizzled-5 / FZD5 Protein, Fc Tag
Catalog No. 502106646
Encompass_Preferred
Recombinant Protein;1MG;Human Frizzled-5, Fc Tag (FZ5-H5259) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Pro 167 (Accession # Q13467).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 42.6 kDa. The protein migrates as 50-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6646 Supplier ACROBiosystems Supplier No. FZ5H52591MG
Recombinant Protein;1MG;Human Frizzled-5, Fc Tag (FZ5-H5259) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Pro 167 (Accession # Q13467).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 42.6 kDa. The protein migrates as 50-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.