Learn More
Sino Biological Human Frizzled-5 / FZD5 Protein (His Tag) 1mg
Supplier:  Sino Biological 10473H08H1
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Pro 167) of human frizzled 5 (NP_003459.2) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6591