Learn More
Sino Biological Recombinant Human Frizzled 6 Protein (His Tag) 1
Supplier:  Sino Biological 16004H08H1
Encompass_Preferred
A DNA sequence encoding the human FZD6 (NP_003497.2) (Met1-Val153) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6806