Learn More
Sino Biological Recombinant Human Chorionic Gonadotropin/CGA Protein (mFc Tag)
Supplier:  Sino Biological 10903H05H50
Encompass_Preferred
A DNA sequence encoding the human CGA (NP_000726.1) (Met1-Ser116) was expressed with the Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-224-6761