Learn More
Sino Biological Human FSHB PAb 20ul (Sino Biological)
Supplier:  Sino Biological 11935T1620
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human FSH Beta / FSHB (rh FSH Beta / FSHB; Catalog#11935-H08H; NP_000501.1; Met1-Glu129). FSH Beta / FSHB specific IgG was purified by Human FSH Beta / FSHB affinity chromatography.
Catalog No. 50-237-0729