Learn More
BIOHIPPO Human FSP Microsample
SDP
Catalog No. 5000313779
Encompass
Human FSP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of FSP in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 068 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human FSP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human FSP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human FSP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13779 Supplier BIOHIPPO Supplier No. BHE15212374
Human FSP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of FSP in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 068 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human FSP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human FSP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human FSP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color