Learn More
Sino Biological Recombinant Human FTCD Protein (His Tag)
Supplier:  Sino Biological 21909H08B100
Encompass_Preferred
A DNA sequence encoding the human FTCD (NP_006648.1) (Met1-Glu541) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-3859