Learn More
Sino Biological Human FTL ELISA Pair Set 15PP (Sino Biological)
Catalog No. 502372013
Encompass_Preferred
Each vial contains 20 ng of recombinant Human FTL/ferritin, light polypeptide. Reconstitute with 1 mL detection antibody dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 3000 pg/mL is recommended.
Catalog No. 50-237-2013 Supplier Sino Biological Supplier No. SEK1248215
Each vial contains 20 ng of recombinant Human FTL/ferritin, light polypeptide. Reconstitute with 1 mL detection antibody dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 3000 pg/mL is recommended.