Learn More
Sino Biological Anti-Ferritin light chain Antibody, Mouse Monoclonal
Supplier:  Sino Biological 12482MM0920
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human FTL / ferritin, light polypeptide (rh FTL / ferritin, light polypeptide; Catalog#12482-H07E; P02792; Met 1-Asp 175). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-8547