Learn More
Sino Biological Anti-Ferritin light chain Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 12482T54100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Ferritin light chain (rh Ferritin light chain; Catalog#12482-H07E; P02792; Met1-Asp175). Ferritin light chain specific IgG was purified by Human Ferritin light chain affinity chromatography.
Catalog No. 50-255-4369