Learn More
Sino Biological Anti-Ferritin light chain Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 12482T1620
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human FTL / ferritin, light polypeptide (rh FTL / ferritin, light polypeptide; Catalog#12482-H07E; P02792; Met1-Asp175). FTL / ferritin, light polypeptide specific IgG was purified by Human FTL / ferritin, light polypeptide affinity chromatography.
Catalog No. 50-241-0530