Learn More
BIOHIPPO Human FTO Microsample
SDP
Catalog No. 5000315355
Encompass
Human FTO Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of FTO in serum plasma and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 059 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human FTO Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human FTO Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human FTO biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15355 Supplier BIOHIPPO Supplier No. BHE15210530
Human FTO Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of FTO in serum plasma and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 059 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human FTO Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human FTO Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human FTO biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color