Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Furin Recombinant Protein

Catalog No. RP062 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP062 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP062 Supplier Invitrogen™ Supplier No. RP062
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

RP-062 has an activity of 1 U/ul, where 1 U (a unit of activity) is defined as the amount of furin that can release 1 pmol of AMC from the fluorogenic peptide pERTKR-MCA in 1 min. As a fully functional enzyme, RP-062 can also be used in a variety of enzymatic assays. 1 U (a unit of activity) is defined as the amount of furin that can release 1 pmol of AMC from the fluorogenic peptide pERTKR-MCA in one minute (1 pmol of AMC/min). This furin construct represents amino acid residues 1-713 from human furin convertase protein.

Furin is a membrane-associated, calcium-dependent, serine protease that belongs to the subtilisin-like prohormone convertase (PC) family. Members of this family of cellular enzymes cleave most prohormones and neuropeptide precursors. Numerous other cellular proteins, some viral proteins, and bacterial toxins that are transported by the constitutive secretory pathway are also targeted for maturation by PCs. Furin and other PC family members share structural similarities which include a heterogeneous ~10 kDa amino-terminal proregion, a highly conserved ~55 kDa subtilisin-like catalytic domain, and carboxyl-terminal domain that is heterogeneous in length and sequence. These enzymes become catalytically active following proregion cleavage within the appropriate cellular compartment. Furin is the only known PC to possess a transmembrane domain. Cleavage of target proteins occurs at the carboxyl-terminus of the furin consensus sequence, RX(K/R)R. It has been shown that the acidic peptide sequence, C771PSDSEEDEG780, localizes furin to the trans-Golgi-network. Phosphorylation of serine residues within this region modulates intracellular routing of furin protein.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Bioactivity, Western Blot
Formulation 0.02M Bis-tris with 1mg/mL BSA and no preservative
Name Human Furin
Purification Method Purified
Quantity 50 μL
Source Non-Human Primate
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Common Name Furin Convertase
Biological Activity Specific Activity = 1 U/ μL; where 1 U is defined as the amount of furin that can release 1 pmol of AMC from the fluorogenic peptide pERTKR-MCA in 1 min.
Product Type Protein
Conjugate Unconjugated
Species Non-human Primate
Recombinant Recombinant
Sequence Human Furin convertase protein, amino acids 1-713
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Activity Specific Activity = 1 U/μL; where 1 U is defined as the amount of furin that can release 1 pmol of AMC from the fluorogenic peptide pERTKR-MCA in 1 min.
Expression System Non-Human Primate cells
Form Liquid
Protein Subtype Recombinant
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.