Learn More
ACROBiosystems Human Furin / PCSK3 Protein, His Tag
Catalog No. 502055058
Encompass_Preferred
Recombinant Protein;20UG;Human Furin, His Tag (FUN-H52H3) is expressed from human 293 cells (HEK293). It contains AA Asp 108 - Glu 715 (Accession # P09958).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 67.5 kDa .
Catalog No. 50-205-5058 Supplier ACROBiosystems Supplier No. FUNH52H320UG
Recombinant Protein;20UG;Human Furin, His Tag (FUN-H52H3) is expressed from human 293 cells (HEK293). It contains AA Asp 108 - Glu 715 (Accession # P09958).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 67.5 kDa .